Order Entry
India
ContactUsLinkComponent
Calibration weights, class 1
Calibration weights, class 1
Catalog # TROE8144W
undefined
Calibration weights, class 1
Catalog # TROE8144W
Specifications
  • Weight:
    100 g
  • Pk:
    1
  • Type:
    Individual weight, Class 1
Specifications
About this item
Precision-crafted of stainless steel with a highly polished finish
  • Weights are straight cylinders
  • Can be used as a reference standard in calibrating other weights
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0,1 mg to 0,01 mg
Delivery information: Supplied with a NVLAP weight calibration certificate.