VWR® Traceable®, Digital thermometers, long-stem
Catalog # VWRU61220-416
Supplier: VWR Collection
Specifications
- Range:–50…+150 °C
- Resolution:0,1 °C
- Weight:<28,3 g
- Length:279 mm
- Environmentally Preferable:
- Accuracy:±1,0 (-20...120) °C
- Pk:1
- Probe:One stainless steel probe
Specifications
About this item
Constructed of ABS plastic and stainless-steel, these thermometers are ideal for replacing all glass and mercury thermometers in the lab. Perfect-sized unit fits into flasks, cuvettes, test tubes, beakers, ovens, and incubators. Read temperatures in foods, gases, samples, reagents, water baths and semisolids.
- On/off switch provides longer battery life, °F/°C switchable range
- Long stem allows for taking readings while stirring
- Readings are updated every second
With stainless steel probe.
Certifications: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Delivery: Supplied with Traceable® certificate, protective sleeve and battery.