Order Entry
India
ContactUsLinkComponent
 
VWR® Redi-Stor™, Conductivity Probe Storage Solution
Catalog # VWRU62344-940
Supplier:  VWR Collection
CAS Number:  
undefined
VWR® Redi-Stor™, Conductivity Probe Storage Solution
Catalog # VWRU62344-940
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Storage solution for conductivity probe, 454 g bottle

Specifications

About this item

Ideal solution for storing conductivity probes. Eliminates probe contamination, preserves probe cleanliness, eliminates growths found when storing in water only, and maintains probe for immediate use with no conditioning.

Delivery: Supplied with step-by-step calibration instructions, individual temperature compensation chart and Traceable® certificate.