VWR®, Algicide for water baths
Catalog # VWRU13272-031
Supplier: VWR Collection
Specifications
- Description:Algicide, 236 ml
- Pk:1
Specifications
About this item
Effective algicide, for use with water in water baths.
- Effective against black algae
- Compatible with stainless steel bath
- Supplied in easy to use plastic dispenser bottle
Use 20 drops of solution per 3,78 litres of water.