VWR® Talon®, Clamp holder
VWRU21572-658
- Spanning width:0 - 19 mm
- Pk:1
Building on the upgraded Q-clamp design, this all-position clamp holder allows adjustment at any angle in any plane.
- Holders are set at 90° to each other connected by a 90° connector for 360° rotation