Order Entry
India
ContactUsLinkComponent
VWR®, Microscope Slides for Printers
Catalog # VWRU89078-508
Supplier:  VWR Collection
undefined
VWR®, Microscope Slides for Printers
Catalog # VWRU89078-508
Supplier:  VWR Collection
Specifications
  • Colour:
    Yellow
  • Description:
    Microscopy slides
  • Pk:
    144
Specifications
About this item
Slides with frosted tab designed to be printed on by inkjet printer for histology, cytology, hematology, and microbiology applications.
Delivery: Frosted tabs are available in various colours.