VWR®, Staining Rack
Catalog # VWRU15153-821
Supplier: VWR Collection
Specifications
- Description:Micro slide staining rack
- Pk:1
Specifications
About this item
Stainless steel.
- Triangular slide with a tapered top
- Contact the slides only in a narrow zone
- Simple and quickly adjustable
- Fit sinks up to 534 mm inside dimensions
Staining rack is designed for use across laboratory sinks of various sizes.