Order Entry
India
ContactUsLinkComponent
VWR® Talon®, Support stand, tripod base
VWR® Talon®, Support stand, tripod base
Catalog # VWRU12985-076
Supplier:  VWR Collection
undefined
VWR® Talon®, Support stand, tripod base
Catalog # VWRU12985-076
Supplier:  VWR Collection
Specifications
  • Description:
    Support stand, tripod base, Ø: 152 mm
  • Pk:
    1
Specifications
About this item
Tripod based support stand for use with VWR® Talon® lab frames.
  • Support stand base is constructed of cast iron with black enamel finish
  • Base features a built-in support rod holder with locking knob that accepts a 1,3 cm (¹/₂") Ø support rod (sold separately)