Order Entry
India
ContactUsLinkComponent
VWR® Talon®, Hook connectors
  VWRU60079-166
undefined
VWR® Talon®, Hook connectors
  VWRU60079-166
  • Spanning width:
    13 mm
  • Description:
    Hook connector
  • Material:
    Nickel-plated zinc
  • Seq:
    02
  • Pk:
    1
This hook connector allows the one-handed assembly of two components with one adjustment screw.
  • Simple, versatile, and easy-to-use
  • Maximum grip size: 13 mm (¹/₂")