VWR® Signature™, Disposable electroporation cuvettes
Catalog # VWRU89047-208
Supplier: VWR Collection
Specifications
- Volume:400 µl
- Description:Electroporation cuvettes with 2 mm gap
- Pk:50
Specifications
About this item
High quality durable polycarbonate has been tested to optimise transformation efficiencies. These cuvettes are built to provide exact tolerances ensuring reproducible results.
- Polished aluminum plates reducing static and arcing
- Compatible with most electroporation devices
- Gamma irradiated to warranty sterility
Packaging: 50 individually wrapped cuvettes.