Order Entry
India
ContactUsLinkComponent
VWR®, Caps for Scintillation Vials
  VWRU66022-140
VWR®, Caps for Scintillation Vials
  VWRU66022-140
  • Thread:
    15-425
  • Description:
    Urea closures with aluminium foil liners, cork-backed
  • Environmentally Preferable:
  • Pk:
    1.000
White, not autoclavable.