VWR®, Water Sampling Bottles, Amber, Boston Type, Round
VWRU89093-934
- Capacity:125 ml
- Description:Amber, quality-assured® (QA), with open-top (22-410)
- Seq:02
- Pk:24
Type III (soda-lime) glass, amber.
- Open-top cap with ultrasonically bonded fluoropolymer resin/silicone septa make them ideal for use in volatile organic analysis
- Amber glass is recommended for light sensitive samples
Quality-Assured® (QA) packers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. Not Processed (NPC) packers do not receive the specialty cleaning process.
: QA packers include certificate of analysis, bar codes with lot number and individual container number for complete traceability.