Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Forceps
VWR®, Tissue forceps
VWR®, Tissue forceps
VWR®, Tissue forceps
VWRU82027-446
:
VWR Collection
VWR®, Tissue forceps
VWRU82027-446
:
VWR Collection
Description:
Tissue forceps, 1×2 teeth
Length:
152 mm
Environmentally Preferable:
Pk:
1
Stainless steel.