Order Entry
India
ContactUsLinkComponent
Anti-static wires for drums
Anti-static wires for drums
Catalog # JUSU08500
undefined
Anti-static wires for drums
Catalog # JUSU08500
Specifications
  • Description:
    Antistatic flexible wire for bonding and grounding, 91,4 cm, with dual alligator clips 16 mm jaws
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
Use antistatic wires to prevent sparks from static electricity generated by movement and flow of flammable liquids.
  • Antistatic wire for bonding and grounding, with dual hand clamps
  • Reduce fire risks by preventing sparks from static electricity
  • Flexible wire
During waste collection operations, use one antistatic wire to bond from a safety container to a receiving vessel such as a funnel, then properly ground by attaching a second antistatic wire to the drum and then to an earth ground. Standard wire lengths are 3-foot (1-M) and 10-foot (3-M); for longer reach, use several wires with ¼-in (6-mm) terminal ends.