VWR®, Inoculating Loops and Needles, Sterile
PS, sterile. Loops and needles are smooth and flexible to facilitate uniform streaking without damaging the gel surface. Needles are straight and suitable for removal of specimens of single colonies.
- Colour coded for easy size identification
- Packaged sterile in safe, tamperproof, zip-seal resealable bags
- Disposable loops and needles do not need flaming, no risk of infection due to improper sterilisation
- Loops and needles can be used under hoods without danger
: Packed sterile in safe, tamperproof, zip-seal resealable bags.
Frequently Bought Together