Order Entry
India
ContactUsLinkComponent
VWR®, Disposal Box for Pipette Tips
VWR®, Disposal Box for Pipette Tips
Catalog # VWRU82010-754
Supplier:  VWR Collection
undefined
VWR®, Disposal Box for Pipette Tips
Catalog # VWRU82010-754
Supplier:  VWR Collection
Specifications
  • Description:
    Disposal Box for Pipette Tips
  • Dimensions:
    152×152×152 mm
  • Pk:
    1
Specifications
About this item
Acrylic, clear.
  • Eliminates any need to handle pipette tips used with hazardous fluids
  • The 10 mm thick acrylic provides protection from beta radiation
  • Large, hinged lid allows easy removal of contents
This box features a flip-top port cover for convenient one-handed disposal of used pipette tips.