VWR®, Flasks, Round Bottom, with Standard Ground Joint, Short Neck
Supplier: VWR Collection
Certificates
About this item
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
The inexpensive alternative for a wide range of laboratory applications. With matt labelling area. Imprinted catalogue number for convenient reordering.
Certifications: According to ASTM Specification E1403, Type II, Class 3 requirements.
Frequently Bought Together