
Tetrabutylammonium hydroxide 0.98-1.02 M in water for HPLC, for ion pair chromatography, J.T.Baker®
Catalog # 9580-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:0.98 - 1.02
- Assay((C₄H₉)₄NOH), Molarity:0.98 - 1.02
- Ultraviolet Absorbance (0.1M Solution in Water) - 240 nm:≤0.15
- Ultraviolet Absorbance (0.1M Solution in Water) - 245 nm:≤0.10
- Ultraviolet Absorbance (0.1M Solution in Water) - 254 nm:≤0.07
- Ultraviolet Absorbance (0.1M Solution in Water) - 280 nm:≤0.04
- Ultraviolet Absorbance (0.1M Solution in Water) - 300 nm:≤0.04
Specification Test Results
About this item
:
:259.48 g/mol
Density:0,83 g/cm³
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:3267
ADR:8,III
:GHS05
CAS Number:2052-49-5
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425