Order Entry
India
ContactUsLinkComponent
 
 
Tetrabutylammonium hydroxide 40% in water, Macron Fine Chemicals™
  E224-80
 :  
undefined
 
Tetrabutylammonium hydroxide 40% in water, Macron Fine Chemicals™
  E224-80
 :  

 

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • For Laboratory,Research,or Manufacturing Use:
    38 - 42 %
  • Assay ((CH₃CH₂CH₂CH₂)₄NOH):
    38 - 42 %

 

 

 :
 :259.48 g/mol
 :Refrigerator
 :218-147-6
 :3267
 :8,II
 :GHS05
 :2052-49-5
 :C₁₆H₃₇NO
 :MFCD00009425