Sodium hydroxide 0.01 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
:Management of Change (MOC) category = NI
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:0.0095 - 0.0105
- Protect from air to avoid absorption of carbon dioxide.:= 5 ppm
- Normality:0.0095 - 0.0105
- Chloride (Cl):≤5 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.5 ppm
Specification Test Results
Frequently Bought Together