Zinc ≥99.8% (by EDTA titration), sticks, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 4274-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:0.1 ppm max.
- For Laboratory,Research,or Manufacturing Use:0.01% max.
- Assay (Zn) (by EDTA titrn):≥99.8 %
- Iron (Fe):≤0.01 %
- Lead (Pb):≤0.01 %
- Suitability for Arsenic Determination:Passes Test
- Trace Impurities - Arsenic (As):≤0.100 ppm
Specification Test Results
About this item
:
:65.39 g/mol
Merck Index:12,10255
Boiling Pt:908 °C (1013 hPa)
Melting Pt:420 °C
Density:7,14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291