Order Entry
India
Orders LinkContactUsLinkComponent
VWR®, Sample library tubes
VWR®, Sample library tubes
Supplier:   VWR Collection
VWR®, Sample library tubes
Supplier:  VWR Collection
Certificates
VWR®, Sample library tubes
VWR®, Sample library tubes
3 Options Available Below
Product Details & Documents
PP, clear.
  • Autoclavable and freezable (–80 °C)
  • Printed with an alpha numeric grid that helps to find a sample easily
These 1,2 ml sample library tubes are supplied in a 96 tube format for storage of specimens. The rack is designed to meet SBS standards and can be used in robotic applications.
Delivery information: Supplied in 8×12 racks with clear, snug fit lid.
Product Family Options
Tubes in racks
Product Information
  • Colour
    Availability
    Price
  • VWRU82006-698
    Individual tubes
    Individual tubes
    Clear
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
  • VWRU93000-040
    Individual tubes, sterile
    Individual tubes, sterile
    Clear
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
  • VWRU82006-700
    8-well tube strips
    8-well tube strips
    Clear
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • 12-well closure strips, sterile
    12-well closure strips, sterile
    VWRU93000-048
    Log in for availability
    1 * 960 items