VWR®, Cleanser, spray, 70% isopropanol, filtered
Consisting of 70% v/v isopropanol blended with highly purified water, this ready to use formulation eliminates mixing and filtration. Used for wiping gloved hands and also as surface cleaner for removal of contaminants particularly when used in conjunction with sterile cleanroom wipers.
- Filtered to 0,2 µm
- Suitable for hard surfaces of cleanrooms, isolators, and laminar flow cabinets
Frequently Bought Together