VWR® Traceable® One-Shot™, Conductivity standards (CRM)
Supplier: VWR Collection
Certificates
About this item
Traceable® Conductivity Standards are among the most accurate available. They are compatible with all makes of equipment, and can be used in lab or field. Each bottle is labeled for calibrating conductivity (microsiemens/micromhos), resistivity (ohms), and dissolved solids (parts per million).
- Certified reference material
- 100% compatible with all makes of instruments and probes
- Calibration within disposable container eliminates contamination
Certifications: Accredited ISO Guide 34, ISO Guide 35, ISO Guide 31, ISO/IEC 17025.
Delivery: Supplied with 480 ml container, step-by-step calibration instructions, individual temperature compensation charts, traceability information and Traceable® certificate.
Frequently Bought Together