VWR®Traceable®, Digital timer, bench-top
Digital timer, bench-top, 0,001 %, 106,4×108×44,5 mm
Catalog # VWRU61161-326
Supplier: VWR Collection
Jump to:
Product Details & Documents
This sturdy ABS timer with easy-to-use over-sized buttons provides precise timing for enzyme tests, viscosity measurements, animal studies, and all timed laboratory tests.
- Timing capacity: 0,01 seconds to 9 hours, 59 minutes, 59 seconds, and 99 hundredths of a second
- Quartz-crystal timing assures a resolution of 0,01 seconds and an accuracy of 0,001%
- Unit switches between cumulative and interval split and allows an infinite number of timeouts
- Three 4 mm sockets (and supplied test leads) allow external triggering of the start/stop and start/lap/reset functions of the timer
- Digital display is 1,9 cm (³/₄") high and is readable from ten feet
Weight: 198,4 g
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Delivery information: Supplied with AA Alkaline battery and Traceable® certificate.
Specifications
- Description:Digital timer, bench-top
- W×D×H:106,4×108×44,5 mm
- Accuracy:0,001 %



