Order Entry
India
ContactUsLinkComponent
VWR® Traceable®Mini-Dolly® , Folding cart
VWR® Traceable®Mini-Dolly® , Folding cart
Catalog # VWRU61161-316
Supplier:  VWR Collection
undefined
VWR® Traceable®Mini-Dolly® , Folding cart
Catalog # VWRU61161-316
Supplier:  VWR Collection

Specifications

  • Description:
    Mini-Dolly folding cart
  • W×D×H:
    648×451×876 mm
  • Pk:
    1
  • Weight:
    9,07 kg

Specifications

About this item

This lightweight platform cart ideal for moving chemicals, carboys, computer paper, filing boxes, and scientific equipment. The cart folds to a compact size for easy storage and features a nonskid 45×64,8 cm platform. Plastic guards on all four sides protect walls, and elastic cords supplied with the cart are designed with quick hooks to help secure the load.

  • Cart folds to a compact-size making it easy to store
  • Super-glide wheels swivel for complete maneuverability
  • Comfortable easy-grip handle extends to 87,6 cm, ensuring a firm hold
  • Eliminates lifting required by high-shelf carts and holds loads up to 120 kg

High-impact ABS plastic with chrome-plated steel.