Order Entry
India
ContactUsLinkComponent
VWR®, UV Crosslinker
VWR®, UV Crosslinker
Catalog # VWRU89131-486
Supplier:  VWR Collection
CAS Number:  
undefined
VWR®, UV Crosslinker
Catalog # VWRU89131-486
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    1
  • Wavelength:
    254 nm
  • Description:
    UV crosslinker, 230 V
  • Ext. W×D×H:
    230×445×280 mm
  • Int. W×D×H:
    287×290×91 mm

Specifications

About this item

VWR® Crosslinker with overhead 254 nm UV is used not only for attaching nucleic acids to a membrane, but also for sterilisation and decontamination of plates, small tools, etc. The Crosslinker assures consistent UV output for many applications including DNA bonding.

  • Preset and manual controls for ultraviolet or time exposures to enable flexibility and reliability of settings
  • Microprocessor that optimizes the use of energy while ensuring consistent, maximum energy efficiency and output
  • Factory presettings that deliver 120000 microjoules or five minutes of exposure and maximum energy setting of 999900 microjoules per cm²
  • Large LED display
  • Door window that blocks UV radiation, but still allows observation of the process
  • Pull out drawer for easy sample placement
  • Five 8 W ultraviolet tubes for uniform overhead UV illumination