Order Entry
India
ContactUsLinkComponent
VWR® Traceable®, Refrigerator and Freezer Plus™ thermometers
VWR® Traceable®, Refrigerator and Freezer Plus™ thermometers
Catalog # VWRU12777-838
Supplier:  VWR Collection
undefined
VWR® Traceable®, Refrigerator and Freezer Plus™ thermometers
Catalog # VWRU12777-838
Supplier:  VWR Collection

Specifications

  • Range:
    –50…+70 °C
  • Resolution:
    0,1 °C
  • Weight:
    113,4 g
  • W×D×H:
    82,5×63,5×16 mm
  • Accuracy:
    ±0,5 °C
  • Pk:
    1
  • Probe:
    One bottled probe

Specifications

About this item

Mercury-free, solid-state bottle probe in nontoxic glycol with three meter cable. These digital thermometers are designed for monitoring temperatures in freezers, water baths, heating blocks, incubators and refrigerators with this enclosed temperature-buffered sensor.

  • Monitors high and low readings overnight or for any time period
  • Programmable alarm when temperature rises above or falls below a set point
  • Triple display simultaneously shows MIN/MAX and current temperatures

Bottle insulates sensor from rapid temperature changes when refrigerator door is opened. Bottle probes are used in monitoring storage conditions for vaccines or other important samples.

Certifications: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: Supplied with battery, Velcro®, magnetic strips and Traceable® certificate.