VWR®, Wash Bottles, Wide Neck, with Coloured Caps, Labeled, Safety-Vented
Supplier: VWR Collection
Certificates
About this item
LDPE, with PP spary cap.
- Unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know safety-labelled wash bottles
- Allows dispensing with bottle upright or inverted
- Volatiles are vented to eliminate solvent drips
- Pre-printed with the chemical name and formula, the National Fire Protection Association (NFPA) Section 704 four-colour diamond, health hazards, Chemical Abstract System (CAS) number and suggested protective clothing and equipment
Labelled with six of the most commonly used solvent names or label as per the need. Solvents compatible with LDPE can be used. Custom labelling is also available.
Certifications: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
Frequently Bought Together