VWR®, Wash Bottles, Narrow or Wide Neck, with Cap
Catalog # VWRU16650-537
Supplier: VWR Collection
Specifications
- Cap Size:53 mm
- Capacity:500 ml
- Seq:02
- Environmentally Preferable:
- Pk:6
Specifications
About this item
Bottles, LDPE red and caps, PP, red, are vividly coloured to aid in the identification of hazardous contents.
- Good chemical resistance
- Good temperature resistance