VWR®, Tissue Processing/Embedding Cassettes, Histology
Histological tissue cassettes, Green
VWRU18000-136
These histology cassettes are ideal for routine tissue processing and embedding processes.
- Resistant to solvents and decalcifying solutions
- Features flow-thru slots
- 10% increased chamber length for maximisation of specimen range
- Textured writing surface for sample identification and tracking
- With 45° print surface is compatible with all automatic printing devices
- Attached covers use frangible hinge
- Colour:Green
- Description:Histological tissue cassettes
- Seq:01



