Order Entry
India
ContactUsLinkComponent
VWR®, Beakers, Low Form, with Handle
VWR®, Beakers, Low Form, with Handle
Catalog # VWRU13916-795
Supplier:  VWR Collection
Supplier #: 13916-795
undefined
VWR®, Beakers, Low Form, with Handle
Catalog # VWRU13916-795
Supplier:  VWR Collection
Supplier #: 13916-795
Log in for availability
Select an Option
100 - 3000 ml
Select an Option
3000 ml
Select an Option
04
Select an Option
1
About this item
PP, translucent.
  • Robust with non-drip spout and ridged thumb-grip on handle
  • Low form for better stability
  • Autoclavable
Frequently Bought Together