VWR®, Hydrometers
These hydrometers are designed for industries such as food and beverage, chemicals, pharmacology, transportation and refrigeration.
- Specific gravity of petroleum products, heavy and light liquids
- Lead free metal ballast
- Corrections to one tenth of the smallest division
: Traceable to NIST and DAkkS. Calibration conforms with A2LA accreditions. ASTM calibrated.
: Supplied with calibration report.
Frequently Bought Together