Order Entry
India
Orders LinkContactUsLinkComponent
VWR®, Microscope slide boxes
VWR®, Microscope slide boxes
Supplier:   VWR Collection
VWR®, Microscope slide boxes
Supplier:  VWR Collection
Certificates
VWR®, Microscope slide boxes
VWR®, Microscope slide boxes
5 Options Available Below
Product Details & Documents
These boxes made of PP are designed for the storage or transport of either 25 or 100 microscopy slides.
  • Boxes are unlined
  • Hinged lids are held securely closed by latch locks
  • Colour-coded for ease in sorting and selection
Convenient, space-saving design with stacking feet maximises stability when boxes are stored atop one another.
Product Family Options
Product Information
  • Colour
    Material
    No. of slides
    W×D×H
    Availability
    Price
  • White
    Plastic
    25
    141×88×35 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • Blue
    Plastic
    25
    141×88×35 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • Red
    Plastic
    25
    141×88×35 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • White
    Plastic
    100
    208×175×34 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • Red
    Plastic
    100
    208×175×34 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items