Order Entry
India
Orders LinkContactUsLinkComponent
 

Adhesion slides

Adhesion slides with 45° clipped corners, White, 75×25 mm
  631-1109
Adhesion slides
  631-1109
Adhesion slides
Adhesion slides

Adhesion slides with 45° clipped corners, White, 75×25 mm

 
Rs.3,969.00
   
1 * 100 items
 

These high quality slides are suitable for automated research and clinical applications.

  • High adhesion to reduce tissue loss and the number of tests
  • Background staining reduced in special dyes
  • Frosted ends for thermal slide printers - Clear and consistent printing
  • Fine glass with high flatness and transmittance for less intrinsic autofluorescence
  • Reduced surface leaching and increased stability and longevity of the slides

Suitable for use in anatomo-cytopathology and academic research laboratories.

 
  • W×D:
    75×25 mm
  • Colour:
    White
  • Description:
    Adhesion slides with 45° clipped corners
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.