Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 3 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® Excel® SuperC18 columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.
- Ultra-inert for maximum performance and reproducibility
- Ultra low bleed for improved LC/MS compatibility
- Wide range of particle sizes (1,7 to 10 µm) and complementary phases
Maximum recommended pH: 1,5 to 11,5
Max. pressure: 1000 bar
Specifications
- Catalog No:
- EXL-1111-1002U
- EXL-1111-2546U
- EXL-1111-1003U
- EXL-1111-0302U
- EXL-1111-7546U
- EXL-1111-1046U
- EXL-1111-1503U
- EXL-1111-1546U
- EXL-1111-7502U
- EXL-1111-1246U
- EXL-1111-1502U
- EXL-1111-0246U
- EXL-1111-0503U
- EXL-1111-0502U
- EXL-1111-2502U
- EXL-1111-0546U
- EXL-1111-2503U
- EXL-1111-1203U
- EXL-1111-2046U
- EXL-1111-0202U
- EXL-1111-0203U
- Carbon load:
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- pH range:
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- Surface area:
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
Specifications
Guard holders
Product Information
Frequently Bought Together