Order Entry
India
ContactUsLinkComponent
VWR®, Spreaders, L-Shaped, Disposable
  89042-021
undefined
VWR®, Spreaders, L-Shaped, Disposable
  89042-021
  • Colour:
    Blue
  • Description:
    Spreaders, L-shaped
  • Seq:
    01
  • Pk:
    100
PP, sterile, disposable L-shaped cell spreaders are ideal for spreading bacterial cultures evenly.
  • L-shape allows application across the entire Petri dish or plate surface
  • Smooth spreading surface with slight upward return reduces the risk of damaging the agar sample
  • Spreaders are presterilised, eliminating the need for flaming or autoclaving
 : Packaged in resealable bags of 10.