Order Entry
India
ContactUsLinkComponent
VWR®, Ready PCR Mix, 2X
  N806-2X1.25ML
   Safer Human and Environmental Health
 :  
undefined
VWR®, Ready PCR Mix, 2X
  N806-2X1.25ML
   Safer Human and Environmental Health
 :  
  • Pk:
    1 KIT
  • Description:
    Ready PCR Mix, 2X, 2×1,25 ml
  • Environmentally Preferable:
    Safer Human and Environmental Health
Ready PCR Mix, 2X offers a single-step procedure for performing PCR reactions followed by analysis on agarose gels without the addition of loading buffer. All components for assembly and performance of PCR reactions as well as loading and visualisation of PCR products on agarose gels are included. The user supplies primer and template DNA.
  • Direct to gel loading
  • Immediate visualisation
  • Safe to use