Order Entry
India
ContactUsLinkComponent
Allihn condensers with full length water cooled joints, KIMBLE®
  KIML431500-2430
undefined
Allihn condensers with full length water cooled joints, KIMBLE®
  KIML431500-2430
  • Cone size:
    24/40 NS
  • Jacket length:
    325 mm
  • Socket size:
    24/40 NS
  • Pk:
    1
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for use in refluxing operations, including soxhlet extractions. It has a straight outlet tube at the top and a water-cooled standard taper inner drip joint at the bottom.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.