VWR® SuperClear, Microcentrifuge tubes, 0,5/1,5/2,0 ml
Ultra-clear PP resin microtubes, graduated.
- Can withstand centrifugation upto 20000×g
- Caps include silicone O-rings for secure storage
- Sterile tubes are sterilised by gamma irrardiation
- Caps have smooth surface for applying labels
- Sustainable design based on virgin resins
: Non sterile microtubes and caps are packed in resealable bags.
Sterile microtubes with attached caps are in sterile bags of 50 tubes, 10 bags/pack.
Sterile microtubes with attached caps are in sterile bags of 50 tubes, 10 bags/pack.
Frequently Bought Together