Order Entry
India
ContactUsLinkComponent
Goggles, V80 MONOGOGGLE* 211, JACKSON SAFETY*
  KIMB14387
undefined
Goggles, V80 MONOGOGGLE* 211, JACKSON SAFETY*
  KIMB14387
  • Type:
    Anti-splash
  • Colour:
    Green
  • Lenses:
    Clear VISICLEAR*
  • Pk:
    1
The V80 monogoggles consist of a polycarbonate lens, a PVC frame, and a neoprene headband.
  • Patented indirect ventilation system provides increased airflow, minimised condensation and fogging and protects against splashes
  • Soft-frame design with pantascopic angle to improve work area visibility
  • Soft, pliable plastic nosepiece for snug, comfortable seal
It can be worn over most prescription glasses.
 : Meets ANSI Z87.1+standards