Order Entry
India
ContactUsLinkComponent
Safety spectacles, pheos guard
Safety spectacles, pheos guard
Catalog # 111-1643
Supplier:  UVEX
undefined
Safety spectacles, pheos guard
Catalog # 111-1643
Supplier:  UVEX
Specifications
  • Type:
    Standard; anti-UV (2C-1,2), anti-scratch, permanent anti-fog
  • Colour:
    Black
  • Seq:
    01
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
The innovative Pheos Guard system is a functional expansion of the Pheos range. It is a combination of Pheos safety glasses, an extra frame and a headband.
  • Comfortable frame creates a close seal to protect from particle ingress and splashes
  • Headband can be added: Ideal for working at heights or in confined spaces
  • Compatible with other PPE
  • Pheos Guard S model is specially designed for narrower heads
Ideal for working environments with dust exposure, due to the combination of anti-fog coated lenses and extra frame.
Certifications: EN 166, EN 170