Manganese reagent set, LR
HACH2243300
: HACH
:
- Chemical method:PAN
- Pk:100 Tests
- Description:Manganese reagent set
- Range:0,006 - 0,700 mg/l Mn
Reagent set used for determination of manganese by the PAN method.
: Reagent set includes ascorbic acid powder pillows, alkaline cyanide reagent and PAN indicator solution (0,1% for 100 tests).