Stoppers, hollow, LDPE
Supplier: HACH
Certificates
About this item
Ideally can be used as a small beaker, sample cup for pH meters, flame photometers and automatic analysers.
- LDPE
- Interchangeable with rubber stoppers
- Safer and economical
- Fits snugly
Frequently Bought Together