Goggles, classic™ 9305
Closed vent; anti-fog, Clear PC, Transparent
DALLS364
These Classic™ goggles has soft, extra-wide frames fit snugly and comfortably around face even over glasses. The wide neoprene strap holds goggles securely in place.
- Fits over most prescription eyewear
- Frosted browguard reduces overhead glare
- Excellent splash protection
- Uvextreme® anti-fog coating minimises fogging in extreme environments
Weight: 105 g
: Meets ANSI Z87.2010; SA Z94.3 standards.
- Type:Closed vent; anti-fog
- Colour:Transparent
- Lenses:Clear PC