Diphosphorus pentoxide ≥99.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
9374-01
: Avantor
:
- Pack type:Glass bottle
- Pk:500 g
- Seq:3645
- For Laboratory,Research,or Manufacturing Use:≥99.0%
- Assay (P₂O₅):≥99.0 %
- Insoluble Matter:≤0.01 %
- Phosphorus Trioxide (P₂O₃):Passes Test
- Ammonium (NH₄):≤0.005 %
- Heavy Metals (as Pb):≤0.005 %
:
:141.94 g/mol
:13,07441
:562 °C
:2,253 g/cm³ (20 °C)
:Ambient
:215-236-1
:1807
:8,II
:GHS05
:1314-56-3
:P₂O₅
:MFCD00011440