Order Entry
India
ContactUsLinkComponent
Sulphuric acid 95.0-98.0%, BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Sulphuric acid 95.0-98.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Sulphuric acid 95.0-98.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    95.0 - 98.0 %
  • Assay (H₂SO₄):
    95.0 - 98.0 %
  • Appearance:
    Passes Test
  • Color (APHA):
    ≤10
  • Specific Gravity at 60°/60°F:
    ≥1.84
  • Residue after Ignition:
    ≤4 ppm
  • Ammonium (NH₄):
    ≤1 ppm
  • Trace Impurities - Arsenic (As):
    ≤0.004 ppm
  • Chloride (Cl):
    ≤0.1 ppm
  • Nitrate (NO₃):
    ≤0.5 ppm
  • Trace Impurities - Aluminum (Al):
    ≤200.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤200.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤100.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤100.0 ppb
  • Trace Impurities - Gold (Au):
    ≤100.0 ppb
  • Heavy Metals (as Pb):
    ≤500.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤200.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤200.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤200.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤100.0 ppb
  • Trace Impurities - Mercury (Hg):
    ≤5.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤200.0 ppb
  • Trace Impurities - Potassium (K):
    ≤300.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤300 ppb
  • Trace Impurities - Tin (Sn):
    ≤200.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤200.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤200.0 ppb

Specification Test Results

Frequently Bought Together