Succinic acid ≥99.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0346-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.0%
- For Laboratory,Research,or Manufacturing Use:185.0 - 191.0 °C
- ACS - Assay (HOCOCH₂CH₂COOH):≥ 99.0 %
- ACS - Melting Point:185.0 - 191.0 °C
- ACS - Insoluble Matter:≤ 0.01 %
- ACS - Residue after Ignition:≤ 0.02 %
- ACS - Chloride (Cl):≤ 0.001 %
- ACS - Nitrogen Compounds (as N):≤ 0.001 %
- ACS - Phosphate (PO₄):≤ 0.001 %
- ACS - Sulfate (SO₄):≤ 0.003 %
- ACS - Heavy Metals (as Pb):≤ 5 ppm
- ACS - Iron (Fe):≤ 5 ppm
Specification Test Results
About this item
:
:118.09 g/mol
Merck Index:13,08953
Boiling Pt:235 °C (1013 hPa)
Melting Pt:182 °C
Density:1,56 g/cm³ (20 °C)
Flash Pt:206 °C
Storage Temperature:Ambient
EINECS:203-740-4
UN:3261
ADR:8,III
:Filtered through a 0.2 µm filter.
:GHS07
CAS Number:110-15-6
Formula:HOOCCH₂CH₂COOH
MDL Number:MFCD00002789