Sodium periodate 99.8-100.3% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
: Sodium metaperiodate
:7790-28-5
:
:NaIO₄
:MFCD00003534
:13,08714
:300 °C (1013 hPa)
:175 °C
:3,87 g/cm³ (20 °C)
:Ambient
:232-197-6
:1479
:5.1,II
:Management of Change (MOC) category = N
:213.89 g/mol
:GHS03
:GHS07
:GHS08
- Environmentally Preferable:
- Meets ACS Reagent Chemical Requirements:99.8 - 100.3%
- For Laboratory,Research,or Manufacturing Use:≤0.02%
- Assay (NaIO₄) (by iodometry):99.8 - 100.3 %
- Other Halogens (as Cl):≤0.02 %
- Trace Impurities - Manganese (Mn):≤3.000 ppm
Frequently Bought Together