


Nitric acid 68,0 - 70,0%, AR® ACS, Macron Fine Chemicals™
Catalog # MACR2704-14
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:Passes Test
- For Laboratory,Research,or Manufacturing Use:68.0 - 70.0%
- Appearance:Passes Test
- ACS - Assay (HNO₃):68.0 - 70.0 %
- ACS - Color (APHA):≤10
- ACS - Heavy Metals (as Pb):≤0.2 ppm
- ACS - Residue after Ignition:≤5 ppm
- ACS - Sulfate (SO₄):≤1 ppm
- Trace Impurities - Arsenic (As):≤0.010 ppm
- Chloride (Cl):≤0.5 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
Specification Test Results
About this item
:
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:120,5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS03
:GHS05
:GHS07
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349